Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C18H16orf86
Entrez Gene ID
788008
Gene Name
chromosome 18 C16orf86 homolog
Gene Aliases
-
Chromosome Location
Chr.18: 35220236 - 35222204 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205459.3 NP_001192388.1 1 - 2 209 55

View Full Product Details