Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERI2
Entrez Gene ID
538719
Gene Name
ERI1 exoribonuclease family member 2
Gene Aliases
-
Chromosome Location
Chr.25: 18540516 - 18556531 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205577.1 NP_001192506.1 7 - 8 945 78
GenBank mRNA BC123611.1 - 7 - 8 686 78

View Full Product Details