Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BICDL2
Entrez Gene ID
507073
Gene Name
BICD family like cargo adaptor 2
Gene Aliases
-
Chromosome Location
Chr.25: 2444406 - 2452294 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205599.1 NP_001192528.1 3 - 4 572 83

View Full Product Details