Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C2H2orf80
Entrez Gene ID
539078
Gene Name
chromosome 2 C2orf80 homolog
Gene Aliases
-
Chromosome Location
Chr.2: 96437056 - 96460429 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205626.2 NP_001192555.2 7 - 8 658 106

View Full Product Details