Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIX2
Entrez Gene ID
539492
Gene Name
SIX homeobox 2
Gene Aliases
-
Chromosome Location
Chr.11: 27425278 - 27429527 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205678.2 NP_001192607.1 1 - 2 561 97

View Full Product Details