Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ADRA1D
Entrez Gene ID
539543
Gene Name
adrenoceptor alpha 1D
Gene Aliases
-
Chromosome Location
Chr.13: 50911092 - 50932314 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205684.2 NP_001192613.2 1 - 2 1119 89

View Full Product Details