Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBSN
Entrez Gene ID
511484
Gene Name
rabenosyn, RAB effector
Gene Aliases
ZFYVE20
Chromosome Location
Chr.22: 57158475 - 57203348 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205700.1 NP_001192629.1 5 - 6 729 72

View Full Product Details