Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DTX1
Entrez Gene ID
512873
Gene Name
deltex E3 ubiquitin ligase 1
Gene Aliases
-
Chromosome Location
Chr.17: 61084997 - 61123903 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205734.1 NP_001192663.1 3 - 4 1119 76

View Full Product Details