Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AMMECR1L
Entrez Gene ID
539958
Gene Name
AMMECR1 like
Gene Aliases
-
Chromosome Location
Chr.2: 4601030 - 4621044 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205735.2 NP_001192664.1 5 - 6 988 69

View Full Product Details