Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GCC1
Entrez Gene ID
539985
Gene Name
GRIP and coiled-coil domain containing 1
Gene Aliases
-
Chromosome Location
Chr.4: 91811881 - 91815279 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205744.1 NP_001192673.1 1 - 2 1031 80

View Full Product Details