Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPINDOC
Entrez Gene ID
513662
Gene Name
spindlin interactor and repressor of chromatin binding
Gene Aliases
C29H11orf84
Chromosome Location
Chr.29: 42166781 - 42178300 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205765.1 NP_001192694.1 2 - 3 455 102

View Full Product Details