Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SALL1
Entrez Gene ID
514467
Gene Name
spalt like transcription factor 1
Gene Aliases
-
Chromosome Location
Chr.18: 19545659 - 19561476 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205790.1 NP_001192719.1 1 - 2 74 69

View Full Product Details