Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D25
Entrez Gene ID
515568
Gene Name
TBC1 domain family member 25
Gene Aliases
-
Chromosome Location
Chr.X: 86664669 - 86678020 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205813.1 NP_001192742.1 2 - 3 361 60

View Full Product Details