Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLIP3
Entrez Gene ID
522965
Gene Name
CAP-Gly domain containing linker protein 3
Gene Aliases
-
Chromosome Location
Chr.18: 46688374 - 46701516 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205898.2 NP_001192827.1 7 - 8 1110 100

View Full Product Details