Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GJB5
Entrez Gene ID
524030
Gene Name
gap junction protein beta 5
Gene Aliases
-
Chromosome Location
Chr.3: 110930965 - 110948284 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205907.2 NP_001192836.1 1 - 2 123 83

View Full Product Details