Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ESPNL
Entrez Gene ID
524380
Gene Name
espin like
Gene Aliases
-
Chromosome Location
Chr.3: 117440975 - 117465058 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205909.2 NP_001192838.2 3 - 4 672 74

View Full Product Details