Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SHC4
Entrez Gene ID
524734
Gene Name
SHC adaptor protein 4
Gene Aliases
-
Chromosome Location
Chr.10: 61284094 - 61420825 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205912.1 NP_001192841.1 8 - 9 1241 61

View Full Product Details