Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XKRX
Entrez Gene ID
524975
Gene Name
XK related X-linked
Gene Aliases
-
Chromosome Location
Chr.X: 51241608 - 51253455 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205914.1 NP_001192843.1 1 - 2 337 73

View Full Product Details