Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATF7IP2
Entrez Gene ID
529852
Gene Name
activating transcription factor 7 interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.25: 9153091 - 9229863 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205967.1 NP_001192896.1 4 - 5 1211 103

View Full Product Details