Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLGN1
Entrez Gene ID
530042
Gene Name
neuroligin 1
Gene Aliases
-
Chromosome Location
Chr.1: 93145361 - 94102525 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205973.2 NP_001192902.1 4 - 5 1290 70
GenBank mRNA BC122827.1 - 4 - 5 1290 70

View Full Product Details