Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLF3
Entrez Gene ID
530358
Gene Name
Kruppel like factor 3
Gene Aliases
-
Chromosome Location
Chr.6: 57942693 - 57978356 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205978.1 NP_001192907.1 4 - 5 886 77

View Full Product Details