Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MICU3
Entrez Gene ID
536426
Gene Name
mitochondrial calcium uptake family member 3
Gene Aliases
EFHA2
Chromosome Location
Chr.27: 20097380 - 20175776 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206097.2 NP_001193026.2 2 - 3 545 126

View Full Product Details