Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NTAN1
Entrez Gene ID
505869
Gene Name
N-terminal asparagine amidase
Gene Aliases
-
Chromosome Location
Chr.25: 13716625 - 13731513 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206144.1 NP_001193073.1 6 - 7 483 92

View Full Product Details