Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS37C
Entrez Gene ID
613817
Gene Name
VPS37C subunit of ESCRT-I
Gene Aliases
-
Chromosome Location
Chr.29: 37450869 - 37481097 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206150.1 NP_001193079.1 2 - 3 154 65

View Full Product Details