Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNIH3
Entrez Gene ID
615236
Gene Name
cornichon family AMPA receptor auxiliary protein 3
Gene Aliases
-
Chromosome Location
Chr.16: 27718419 - 27875794 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206204.2 NP_001193133.1 5 - 6 457 83

View Full Product Details