The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001192453.2 | NP_001179382.1 | 3 - 4 | 304 | 103 |
GenBank mRNA | BC108114.1 | - | 3 - 4 | 267 | 103 |