Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDZD8
Entrez Gene ID
519573
Gene Name
PDZ domain containing 8
Gene Aliases
-
Chromosome Location
Chr.26: 37606719 - 37687376 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192510.3 NP_001179439.2 1 - 2 1120 78

View Full Product Details