Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MKX
Entrez Gene ID
521239
Gene Name
mohawk homeobox
Gene Aliases
-
Chromosome Location
Chr.13: 36823229 - 36894467 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192534.1 NP_001179463.1 1 - 2 128 66

View Full Product Details