Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRAP2
Entrez Gene ID
522346
Gene Name
GRB2 related adaptor protein 2
Gene Aliases
-
Chromosome Location
Chr.5: 111263378 - 111329318 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192560.1 NP_001179489.1 6 - 7 829 71

View Full Product Details