Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BATF2
Entrez Gene ID
522469
Gene Name
basic leucine zipper ATF-like transcription factor 2
Gene Aliases
-
Chromosome Location
Chr.29: 43205620 - 43212754 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192561.2 NP_001179490.1 2 - 3 147 76

View Full Product Details