Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CBLL1
Entrez Gene ID
524139
Gene Name
Cbl proto-oncogene like 1
Gene Aliases
-
Chromosome Location
Chr.4: 48758895 - 48775785 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192592.2 NP_001179521.1 2 - 3 202 105

View Full Product Details