Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIGLEC10
Entrez Gene ID
504258
Gene Name
sialic acid binding Ig like lectin 10
Gene Aliases
-
Chromosome Location
Chr.18: 57462616 - 57470264 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206277.1 NP_001193206.1 8 - 9 1790 71

View Full Product Details