Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TNFRSF12A
Entrez Gene ID
617439
Gene Name
TNF receptor superfamily member 12A
Gene Aliases
Fn14
Chromosome Location
Chr.25: 2435936 - 2439695 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206327.1 NP_001193256.1 3 - 4 360 87
GenBank mRNA BF603409.1 - 3 - 4 355 87

View Full Product Details