Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO40
Entrez Gene ID
613597
Gene Name
F-box protein 40
Gene Aliases
-
Chromosome Location
Chr.1: 66092226 - 66111702 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206393.1 NP_001193322.1 2 - 3 1953 70

View Full Product Details