Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIT1
Entrez Gene ID
512027
Gene Name
signaling threshold regulating transmembrane adaptor 1
Gene Aliases
-
Chromosome Location
Chr.8: 59827540 - 59829096 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206418.1 NP_001193347.1 1 - 2 164 116
GenBank mRNA BC149731.1 - 1 - 2 165 116

View Full Product Details