Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DPPA2
Entrez Gene ID
613891
Gene Name
developmental pluripotency associated 2
Gene Aliases
-
Chromosome Location
Chr.1: 54058687 - 54070616 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206470.1 NP_001193399.1 1 - 2 95 85

View Full Product Details