Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ZNF18
Entrez Gene ID
506638
Gene Name
zinc finger protein 18
Gene Aliases
-
Chromosome Location
Chr.19: 30616167 - 30640042 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206494.1 NP_001193423.1 4 - 5 841 80
GenBank mRNA BC105454.1 - 4 - 5 842 80

View Full Product Details