Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GSG1L
Entrez Gene ID
511049
Gene Name
GSG1 like
Gene Aliases
-
Chromosome Location
Chr.25: 25339159 - 25591739 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192728.1 NP_001179657.1 6 - 7 898 58
GenBank mRNA CO887300.1 - 6 - 7 611 58

View Full Product Details