Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPIN1
Entrez Gene ID
541068
Gene Name
spindlin 1
Gene Aliases
-
Chromosome Location
Chr.8: 89648860 - 89730471 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192821.3 NP_001179750.1 2 - 3 262 73

View Full Product Details