Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SALL4
Entrez Gene ID
541170
Gene Name
spalt like transcription factor 4
Gene Aliases
-
Chromosome Location
Chr.13: 79567373 - 79583584 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192836.3 NP_001179765.3 2 - 3 2485 85

View Full Product Details