Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OVOS2
Entrez Gene ID
508026
Gene Name
ovostatin 2
Gene Aliases
OVOS
Chromosome Location
Chr.5: 100679985 - 100732010 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164024.1 NP_001157496.1 18 - 19 2578 100

View Full Product Details