Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAAP20
Entrez Gene ID
508039
Gene Name
Fanconi anemia core complex associated protein 20
Gene Aliases
C16H1orf86
Chromosome Location
Chr.16: 50593781 - 50598411 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166564.1 NP_001160036.1 1 - 2 110 69
GenBank mRNA BC142525.1 - 1 - 2 86 69
EV685494.1 - 1 - 2 112 69

View Full Product Details