Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NR2E3
Entrez Gene ID
281944
Gene Name
nuclear receptor subfamily 2 group E member 3
Gene Aliases
-
Chromosome Location
Chr.10: 18653771 - 18659405 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167900.1 NP_001161372.1 7 - 8 1152 61

View Full Product Details