Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBMX
Entrez Gene ID
509591
Gene Name
RNA binding motif protein X-linked
Gene Aliases
-
Chromosome Location
Chr.Unknown: 8049333 - 8058157 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172039.1 NP_001165510.1 8 - 9 981 103
GenBank mRNA GU304600.1 - 8 - 9 981 103

View Full Product Details