Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC8B1
Entrez Gene ID
508887
Gene Name
solute carrier family 8 member B1
Gene Aliases
SLC24A6
Chromosome Location
Chr.17: 61286699 - 61316746 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177596.1 NP_001171067.1 11 - 12 1574 60
GenBank mRNA BT026307.1 - 5 - 6 568 60

View Full Product Details