The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177596.1 | NP_001171067.1 | 14 - 15 | 1990 | 92 |
GenBank mRNA | BT026307.1 | - | 8 - 9 | 984 | 92 |