Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MSMB
Entrez Gene ID
616019
Gene Name
microseminoprotein beta
Gene Aliases
-
Chromosome Location
Chr.28: 44061311 - 44071858 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190300.2 NP_001177229.1 1 - 2 136 101
GenBank mRNA BC108152.1 - 1 - 2 137 101

View Full Product Details