Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSPAN19
Entrez Gene ID
781222
Gene Name
tetraspanin 19
Gene Aliases
-
Chromosome Location
Chr.5: 14645230 - 14673204 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191125.4 NP_001178054.1 4 - 5 339 61
GenBank mRNA BC122812.1 - 4 - 5 356 61

View Full Product Details