Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LHFPL1
Entrez Gene ID
781310
Gene Name
LHFPL tetraspan subfamily member 1
Gene Aliases
-
Chromosome Location
Chr.X: 61079935 - 61150832 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191127.1 NP_001178056.1 1 - 2 380 61

View Full Product Details