Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LGSN
Entrez Gene ID
538526
Gene Name
lengsin, lens protein with glutamine synthetase domain
Gene Aliases
GLULD1
Chromosome Location
Chr.2: 315119 - 382536 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191136.3 NP_001178065.1 1 - 2 59 92

View Full Product Details